Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Thiazolecarboxaldehyde, 98%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Glass Ampule with Screw-cap
Glass bottle
10200-59-6
C4H3NOS
113.14
ZGTFNNUASMWGTM-UHFFFAOYSA-N
2-thiazolecarboxaldehyde,thiazole-2-carbaldehyde,2-formylthiazole,thiazole-2-carboxaldehyde,2-formyl thiazole,thiazolecarboxaldehyde,2-formyl-1,3-thiazole,1,3-thiazole-2-carboxaldehyde,2-thiazolecarbaldehyde,pubchem9929
2734903
CHEBI:43623
1,3-thiazole-2-carbaldehyde
C1=CSC(=N1)C=O
10200-59-6
C4H3NOS
113.14
ZGTFNNUASMWGTM-UHFFFAOYSA-N
2-thiazolecarboxaldehyde,thiazole-2-carbaldehyde,2-formylthiazole,thiazole-2-carboxaldehyde,2-formyl thiazole,thiazolecarboxaldehyde,2-formyl-1,3-thiazole,1,3-thiazole-2-carboxaldehyde,2-thiazolecarbaldehyde,pubchem9929
2734903
CHEBI:43623
1,3-thiazole-2-carbaldehyde
C1=CSC(=N1)C=O
Specific Gravity 1.288
Flash Point 68°C
Quantity 1g
Refractive Index 1.5735 to 1.5755
Packaging Glass bottle
Color Yellow to Brown
Infrared Spectrum Authentic
Boiling Point 76.0°C to 78.0°C (24.0mmHg)
Density 1.2880g/mL
Assay Percent Range 97.5% min. (GC)
Solubility Information Solubility in water: soluble.
Formula Weight 113.14
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2-Thiazolecarboxaldehyde
Show More Show Less

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue

GHS Signal Word: Warning