Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Thiazolylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 mL
Packaging:
ChemSeal™ bottles
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
Specifications
Color | Black to Yellow |
Density | 0.992 g/mL |
Packaging | ChemSeal™ bottles |
Quantity | 50 mL |
UN Number | UN3399 |
Sensitivity | Air sensitive |
Formula Weight | 229.4 |
Physical Form | Liquid |
Chemical Name or Material | 2-Thiazolylzinc bromide |
Safety and Handling
Hazard Category | H225-H260-H302-H314-H335-H336-H351 |
Hazard Statement | GHS H Statement H225-H261-H314-H351-H335 Highly flammable liquid and vapor. In contact with water releases flammable gas. Causes severe skin burns and eye damage. Suspected of causing cancer. May cause respiratory irritation. |
Precautionary Statement | P201-P202-P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 4.3; Packaging Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE |
TSCA | No |
storageNote1 | 2° to 8°C |
Recommended Storage | Keep cold; Store under Argon |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction