Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Thiohydantoin, 98%, Acros Organics™

Click to view available options
Quantity:
5g
100g
503-87-7
C3H4N2OS
116.14
UGWULZWUXSCWPX-UHFFFAOYSA-N
2-thiohydantoin,thiohydantoin,4-imidazolidinone, 2-thioxo,2-thioxoimidazolidin-4-one,2-thioguidanthion,hydantoin, 2-thio,usaf be-25,2-thioxo-4-imidazolidinone,2-thioxo-1,3-diazolidin-4-one,glycine thiohydantoin
1274030
2-sulfanylideneimidazolidin-4-one
C1C(=O)NC(=S)N1
503-87-7
C3H4N2OS
116.14
UGWULZWUXSCWPX-UHFFFAOYSA-N
2-thiohydantoin,thiohydantoin,4-imidazolidinone, 2-thioxo,2-thioxoimidazolidin-4-one,2-thioguidanthion,hydantoin, 2-thio,usaf be-25,2-thioxo-4-imidazolidinone,2-thioxo-1,3-diazolidin-4-one,glycine thiohydantoin
1274030
2-sulfanylideneimidazolidin-4-one
C1C(=O)NC(=S)N1
Melting Point 229.0°C to 231.0°C
Infrared Spectrum Authentic
Quantity 5g
Packaging Glass bottle
Assay Percent Range 98%
Beilstein 24,26
Solubility Information Solubility in water: slightly.
Formula Weight 116.14
Percent Purity 98%
Chemical Name or Material 2-Thiohydantoin

GHS H Statement Harmful if swallowed.

GHS P Statement Wear protective gloves/eye protection/face protection. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.

Warning