Alfa Aesar™ 2-Thiohydantoin, 99%

Manufacturer: Alfa Aesar™ B2493414

Quantity 25g

 View more versions of this product

Catalog No. AAB2493414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 503-87-7
    Molecular Formula C3H4N2OS
    Formula Weight 116.14
    MDL Number MFCD00005277
    Chemical Name or Material 2-Thiohydantoin

    Specifications

    Formula Weight 116.14
    TSCA TSCA
    Health Hazard 2 GHS H Statement
    H302
    Harmful if swallowed.
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number MU4200000
    Melting Point 229° to 231°C (dec.)
    Quantity 25g
    Molecular Formula C3H4N2OS
    MDL Number MFCD00005277
    CAS 503-87-7
    Health Hazard 1 Warning
    Chemical Name or Material 2-Thiohydantoin
    Assay 99%

    GHS H Statement
    H302
    Harmful if swallowed.

    GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit