Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Thiopheneacetic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass Bottle
Plastic bottle
1918-77-0
C6H6O2S
142.17
MFCD00005458
SMJRBWINMFUUDS-UHFFFAOYSA-N
2-thiopheneacetic acid,2-thienylacetic acid,thiopheneacetic acid,2-thiophen-2-yl acetic acid,thiophene-2-acetic acid,2-2-thienyl acetic acid,2-thiophene acetic acid,thiophen-2-ylacetic acid,thien-2-yl acetic acid,2-carboxymethyl thiophene
15970
CHEBI:45807
2-thiophen-2-ylacetic acid
C1=CSC(=C1)CC(=O)O
1918-77-0
C6H6O2S
142.17
MFCD00005458
SMJRBWINMFUUDS-UHFFFAOYSA-N
2-thiopheneacetic acid,2-thienylacetic acid,thiopheneacetic acid,2-thiophen-2-yl acetic acid,thiophene-2-acetic acid,2-2-thienyl acetic acid,2-thiophene acetic acid,thiophen-2-ylacetic acid,thien-2-yl acetic acid,2-carboxymethyl thiophene
15970
CHEBI:45807
2-thiophen-2-ylacetic acid
C1=CSC(=C1)CC(=O)O
Boiling Point 160°C (22.0mmHg)
Quantity 5g
Color Brown
Infrared Spectrum Authentic
Packaging Glass Bottle
Melting Point 61°C to 65°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 18, 293
Formula Weight 142.17
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 2-Thiopheneacetic acid, 98%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 217-639-8

TSCA : TSCA