Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Thiopheneboronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
6165-68-0
C4H5BO2S
127.96
ARYHTUPFQTUBBG-UHFFFAOYSA-N
thiophene-2-boronic acid,2-thiopheneboronic acid,2-thienylboronic acid,thienylboronic acid,2-thiopheneboronicacid,thiophen-2-ylboranediol,thiophene2-boronic acid,thiophen-2-yl boronic acid,2-thiophenylboric acid,boronic acid, thienyl
2733960
thiophen-2-ylboronic acid
B(C1=CC=CS1)(O)O
6165-68-0
C4H5BO2S
127.96
ARYHTUPFQTUBBG-UHFFFAOYSA-N
thiophene-2-boronic acid,2-thiopheneboronic acid,2-thienylboronic acid,thienylboronic acid,2-thiopheneboronicacid,thiophen-2-ylboranediol,thiophene2-boronic acid,thiophen-2-yl boronic acid,2-thiophenylboric acid,boronic acid, thienyl
2733960
thiophen-2-ylboronic acid
B(C1=CC=CS1)(O)O
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 1g
Melting Point 120.0°C to 122.0°C
Color White to Beige
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Formula Weight 127.96
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 2-Thiopheneboronic acid
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning