Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Thiophenecarbonyl chloride, 98%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
Packaging:
Glass bottle
5271-67-0
C5H3ClOS
146.6
MFCD00005428
QIQITDHWZYEEPA-UHFFFAOYSA-N
2-thiophenecarbonyl chloride,2-thenoyl chloride,2-thienylcarbonyl chloride,2-thiophenecarbonylchloride,thiophenecarbonyl chloride,2-thiophene carbonyl chloride,thiophene-2-carboxylic acid chloride,thenoyl chloride,thienoyl chloride,2-thenoylchloride
78928
thiophene-2-carbonyl chloride
C1=CSC(=C1)C(=O)Cl
5271-67-0
C5H3ClOS
146.6
MFCD00005428
QIQITDHWZYEEPA-UHFFFAOYSA-N
2-thiophenecarbonyl chloride,2-thenoyl chloride,2-thienylcarbonyl chloride,2-thiophenecarbonylchloride,thiophenecarbonyl chloride,2-thiophene carbonyl chloride,thiophene-2-carboxylic acid chloride,thenoyl chloride,thienoyl chloride,2-thenoylchloride
78928
thiophene-2-carbonyl chloride
C1=CSC(=C1)C(=O)Cl
Specific Gravity 1.37
Flash Point 90°C
Infrared Spectrum Authentic
Boiling Point 205°C to 208°C
Refractive Index 1.5890 to 1.591
Density 1.3700g/mL
Packaging Glass bottle
Quantity 25mL
Color Colorless to Gray-Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 146.6
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2-Thiophenecarbonyl chloride, 98%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 226-092-4

TSCA : TSCA