Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Thiophenecarboxaldehyde 98%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
1L
Packaging:
Glass bottle
98-03-3
C5H4OS
112.15
MFCD00005429
CNUDBTRUORMMPA-UHFFFAOYSA-N
2-thiophenecarboxaldehyde,2-thenaldehyde,thiophene-2-aldehyde,2-formylthiophene,2-thienylaldehyde,2-thiophenealdehyde,thiophene-2-carboxaldehyde,2-thienylcarboxaldehyde,2-thienaldehyde,alpha-formylthiophene
7364
CHEBI:87301
thiophene-2-carbaldehyde
C1=CSC(=C1)C=O
98-03-3
C5H4OS
112.15
MFCD00005429
CNUDBTRUORMMPA-UHFFFAOYSA-N
2-thiophenecarboxaldehyde,2-thenaldehyde,thiophene-2-aldehyde,2-formylthiophene,2-thienylaldehyde,2-thiophenealdehyde,thiophene-2-carboxaldehyde,2-thienylcarboxaldehyde,2-thienaldehyde,alpha-formylthiophene
7364
CHEBI:87301
thiophene-2-carbaldehyde
C1=CSC(=C1)C=O
Refractive Index 1.5890 to 1.5920
Infrared Spectrum Authentic
Specific Gravity 1.2
Flash Point 77°C
Density 1.2000g/mL
Packaging Glass bottle
Color Yellow to Brown
Quantity 1L
Boiling Point 198.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 17,285
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, ether and benzene
Formula Weight 112.15
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2-Thiophenecarboxaldehyde, 98%
Show More Show Less

GHS H Statement:
May cause an allergic skin reaction.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

AIR SENSITIVE

EINECSNumber : 202-629-8

RTECSNumber : XM8135000

TSCA : TSCA

Recommended Storage : Product may darken on standing