Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Thiophenecarboxamide, 99%, Thermo Scientific™

Catalog Number ac305420050
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
5813-89-8
C5H5NOS
127.17
MFCD00010425
DENPQNAWGQXKCU-UHFFFAOYSA-N
2-thiophenecarboxamide, 2-thienylamide, 2-thenamide, thiophen-2-carboxamide, 2-aminocarbonyl thiophene, 2-thienylamide french, thiophenecarboxamide, thiolamide, acmc-1ao1m, 5-18-06-00162 beilstein handbook reference
22063
thiophene-2-carboxamide
C1=CSC(=C1)C(=O)N
5813-89-8
C5H5NOS
127.17
MFCD00010425
DENPQNAWGQXKCU-UHFFFAOYSA-N
2-thiophenecarboxamide, 2-thienylamide, 2-thenamide, thiophen-2-carboxamide, 2-aminocarbonyl thiophene, 2-thienylamide french, thiophenecarboxamide, thiolamide, acmc-1ao1m, 5-18-06-00162 beilstein handbook reference
22063
thiophene-2-carboxamide
C1=CSC(=C1)C(=O)N
CAS Min % 98.5
CAS Max % 100.0
Melting Point 179.0°C to 182.0°C
Color Gray to White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 127.17
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2-Thiophenecarboxamide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit