Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Thiophenecarboxylic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
500g
527-72-0
C5H4O2S
128.145
MFCD00005437
QERYCTSHXKAMIS-UHFFFAOYSA-N
2-thiophenecarboxylic acid,2-thenoic acid,2-carboxythiophene,2-thiophenic acid,2-thienylcarboxylic acid,alpha-thiophenecarboxylic acid,2-tca,thiophene-2-carboxylate,tenoic acid,.alpha.-thiophenecarboxylic acid
10700
CHEBI:71241
thiophene-2-carboxylic acid
C1=CSC(=C1)C(=O)O
527-72-0
C5H4O2S
128.145
MFCD00005437
QERYCTSHXKAMIS-UHFFFAOYSA-N
2-thiophenecarboxylic acid,2-thenoic acid,2-carboxythiophene,2-thiophenic acid,2-thienylcarboxylic acid,alpha-thiophenecarboxylic acid,2-tca,thiophene-2-carboxylate,tenoic acid,.alpha.-thiophenecarboxylic acid
10700
CHEBI:71241
thiophene-2-carboxylic acid
C1=CSC(=C1)C(=O)O
Quantity 100g
Packaging Glass bottle
Melting Point 126.0°C to 130.0°C
Boiling Point 260.0°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Solubility Information Solubility in water: 80g/L (20°C). Other solubilities: soluble in alcohol and ether, moderately soluble in chcl3, slightly soluble in petroleum ether
Formula Weight 128.15
Percent Purity 99%
Chemical Name or Material 2-Thiophenecarboxylic acid
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning