Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Thiophenecarboxylic acid hydrazide, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
2361-27-5
C5H6N2OS
142.18
MFCD00005435
SOGBOGBTIKMGFS-UHFFFAOYSA-N
2-thiophenecarboxylic acid hydrazide,thiophene-2-carboxylic acid hydrazide,2-thenoylhydrazine,2-thiophenecarboxylic hydrazide,2-thiophenecarbohydrazide,thiophene-2-carboxylic hydrazide,2-thiophenecarboxylic acid, hydrazide,2-thiophenecarbohydrazonic acid,2-thienylcarboxylic acid hydrazide,2-thiophene carboxylic hydrazide
73098
thiophene-2-carbohydrazide
C1=CSC(=C1)C(=O)NN
2361-27-5
C5H6N2OS
142.18
MFCD00005435
SOGBOGBTIKMGFS-UHFFFAOYSA-N
2-thiophenecarboxylic acid hydrazide,thiophene-2-carboxylic acid hydrazide,2-thenoylhydrazine,2-thiophenecarboxylic hydrazide,2-thiophenecarbohydrazide,thiophene-2-carboxylic hydrazide,2-thiophenecarboxylic acid, hydrazide,2-thiophenecarbohydrazonic acid,2-thienylcarboxylic acid hydrazide,2-thiophene carboxylic hydrazide
73098
thiophene-2-carbohydrazide
C1=CSC(=C1)C(=O)NN
Melting Point 136.0°C to 139.0°C
Quantity 25g
Infrared Spectrum Authentic
Color Yellow to Beige
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 18, 291
Formula Weight 142.18
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 2-Thiophenecarboxylic acid hydrazide, 97%
Show More Show Less

EINECSNumber : 219-107-0

TSCA : TSCA