Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Thiouracil 98.0+%, TCI America™

Click to view available options
Quantity:
25g
250g
141-90-2
C4H4N2OS
128.149
MFCD00006039
ZEMGGZBWXRYJHK-UHFFFAOYSA-N
2-thiouracil,thiouracil,4-hydroxy-2-mercaptopyrimidine,antagothyroid,antagothyroil,deracil,nobilen,2-thioxo-2,3-dihydropyrimidin-4 1h-one,2-mercaptopyrimidin-4-ol,2-mercapto-4-hydroxypyrimidine
1269845
CHEBI:348530
2-sulfanylidene-1H-pyrimidin-4-one
C1=CNC(=S)NC1=O
141-90-2
C4H4N2OS
128.149
MFCD00006039
ZEMGGZBWXRYJHK-UHFFFAOYSA-N
2-thiouracil,thiouracil,4-hydroxy-2-mercaptopyrimidine,antagothyroid,antagothyroil,deracil,nobilen,2-thioxo-2,3-dihydropyrimidin-4 1h-one,2-mercaptopyrimidin-4-ol,2-mercapto-4-hydroxypyrimidine
1269845
CHEBI:348530
2-sulfanylidene-1H-pyrimidin-4-one
C1=CNC(=S)NC1=O
Quantity 250g
Melting Point 331°C
Color White-Yellow
UN Number 3077
Formula Weight 128.15
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material 2-Thiouracil

EINECSNumber : (5)-0925

RTECSNumber : YR1575000

TSCA : Yes