Alfa Aesar™ 2-(Tri-n-butylstannyl)pyridine, tech. 80%

Manufacturer: Alfa Aesar™ B2491114

Quantity 25g

 View more versions of this product

Catalog No. AAB2491114

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 17997-47-6
    Molecular Formula C17H31NSn
    Formula Weight 368.15
    MDL Number MFCD00052052
    UN Number UN2788
    Beilstein 479329
    Synonym Tri-n-butyl-2-pyridyltin
    Chemical Name or Material 2-(Tri-n-butylstannyl)pyridine
    Grade Technical Grade

    Specifications

    Formula Weight 368.15
    Quantity 25g
    Molecular Formula C17H31NSn
    MDL Number MFCD00052052
    CAS 17997-47-6
    UN Number UN2788
    Beilstein 479329
    Synonym Tri-n-butyl-2-pyridyltin
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H372-H312-H315-H319
    Toxic if swallowed.
    Causes damage to organs through prolonged or repeated exposure.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P260-P280-P301+P310-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Recommended Storage 2° to 8°C
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
    Boiling Point 130° to 132°C (0.8mmHg)
    Chemical Name or Material 2-(Tri-n-butylstannyl)pyridine
    Assay 80%
    Grade Technical Grade

    GHS H Statement
    H301-H372-H312-H315-H319
    Toxic if swallowed.
    Causes damage to organs through prolonged or repeated exposure.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.

    2° to 8°C

    GHS P Statement
    P260-P280-P301+P310-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit