Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α,α,2-Trichloro-6-fluorotoluene, 99%, Acros Organics

Click to view available options
Quantity:
25g
62476-62-4
C7H4Cl3F
213.457
MFCD00000830
LQIXIKRJISFVEC-UHFFFAOYSA-N
1-chloro-2-dichloromethyl-3-fluorobenzene,2-chloro-6-fluorobenzal chloride,a,a,2-trichloro-6-fluorotoluene,alpha,alpha,2-trichloro-6-fluorotoluene,benzene,1-chloro-2-dichloromethyl-3-fluoro,2-dichloromethyl-3-chloro-1-fluorobenzene,1-chloro-2-dichloromethyl-3-fluorobenzene #,benzene, 1-chloro-2-dichloromethyl-3-fluoro
600965
1-chloro-2-(dichloromethyl)-3-fluorobenzene
C1=CC(=C(C(=C1)Cl)C(Cl)Cl)F
62476-62-4
C7H4Cl3F
213.457
MFCD00000830
LQIXIKRJISFVEC-UHFFFAOYSA-N
1-chloro-2-dichloromethyl-3-fluorobenzene,2-chloro-6-fluorobenzal chloride,a,a,2-trichloro-6-fluorotoluene,alpha,alpha,2-trichloro-6-fluorotoluene,benzene,1-chloro-2-dichloromethyl-3-fluoro,2-dichloromethyl-3-chloro-1-fluorobenzene,1-chloro-2-dichloromethyl-3-fluorobenzene #,benzene, 1-chloro-2-dichloromethyl-3-fluoro
600965
1-chloro-2-(dichloromethyl)-3-fluorobenzene
C1=CC(=C(C(=C1)Cl)C(Cl)Cl)F
Color Colorless
Density 1.44g/mL
Refractive Index 1.5500 to 1.5520 (20°C, 589nm)
Quantity 25g
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula ClC6H3(F)CHCl2
Beilstein 05, III, 701
Formula Weight 213.47
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material α, α, 2-Trichloro-6-fluorotoluene, 99%
Show More Show Less