Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Trichloroacetyl)pyrrole, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
35302-72-8
C6H4Cl3NO
212.46
MFCD00128757
BBFDGMDENAEMKF-UHFFFAOYSA-N
2-trichloroacetyl pyrrole,2,2,2-trichloro-1-1h-pyrrol-2-yl ethanone,2,2,2-trichloro-1-1h-pyrrol-2-yl ethan-1-one,2-trichloroacetyl-1h-pyrrole,2-trichloroacetylpyrrole,2,2,2-trichloro-1-1h-pyrrol-2-yl-ethanone,ethanone, 2,2,2-trichloro-1-1h-pyrrol-2-yl,2,2,2-trichloro-1-pyrrol-2-ylethan-1-one,trichloroacetyl pyrrole
321487
2,2,2-trichloro-1-(1H-pyrrol-2-yl)ethanone
C1=CNC(=C1)C(=O)C(Cl)(Cl)Cl
35302-72-8
C6H4Cl3NO
212.46
MFCD00128757
BBFDGMDENAEMKF-UHFFFAOYSA-N
2-trichloroacetyl pyrrole,2,2,2-trichloro-1-1h-pyrrol-2-yl ethanone,2,2,2-trichloro-1-1h-pyrrol-2-yl ethan-1-one,2-trichloroacetyl-1h-pyrrole,2-trichloroacetylpyrrole,2,2,2-trichloro-1-1h-pyrrol-2-yl-ethanone,ethanone, 2,2,2-trichloro-1-1h-pyrrol-2-yl,2,2,2-trichloro-1-pyrrol-2-ylethan-1-one,trichloroacetyl pyrrole
321487
2,2,2-trichloro-1-(1H-pyrrol-2-yl)ethanone
C1=CNC(=C1)C(=O)C(Cl)(Cl)Cl
Color Green
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 50g
Elemental Analysis 33.07 to 34.76% C, 6.43 to 7.75% N
Melting Point 72.0°C to 74.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 212.46
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 2-(Trichloroacetyl)pyrrole
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning