Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(Trichloroacetyl)pyrrole, ≥99%

Click to view available options
Quantity:
5g
25g
35302-72-8
C6H4Cl3NO
212.454
MFCD00128757
BBFDGMDENAEMKF-UHFFFAOYSA-N
2-trichloroacetyl pyrrole,2,2,2-trichloro-1-1h-pyrrol-2-yl ethanone,2,2,2-trichloro-1-1h-pyrrol-2-yl ethan-1-one,2-trichloroacetyl-1h-pyrrole,2-trichloroacetylpyrrole,2,2,2-trichloro-1-1h-pyrrol-2-yl-ethanone,ethanone, 2,2,2-trichloro-1-1h-pyrrol-2-yl,2,2,2-trichloro-1-pyrrol-2-ylethan-1-one,trichloroacetyl pyrrole
321487
2,2,2-trichloro-1-(1H-pyrrol-2-yl)ethanone
C1=CNC(=C1)C(=O)C(Cl)(Cl)Cl
35302-72-8
C6H4Cl3NO
212.454
MFCD00128757
BBFDGMDENAEMKF-UHFFFAOYSA-N
2-trichloroacetyl pyrrole,2,2,2-trichloro-1-1h-pyrrol-2-yl ethanone,2,2,2-trichloro-1-1h-pyrrol-2-yl ethan-1-one,2-trichloroacetyl-1h-pyrrole,2-trichloroacetylpyrrole,2,2,2-trichloro-1-1h-pyrrol-2-yl-ethanone,ethanone, 2,2,2-trichloro-1-1h-pyrrol-2-yl,2,2,2-trichloro-1-pyrrol-2-ylethan-1-one,trichloroacetyl pyrrole
321487
2,2,2-trichloro-1-(1H-pyrrol-2-yl)ethanone
C1=CNC(=C1)C(=O)C(Cl)(Cl)Cl
Quantity 25g
Color Gray
Melting Point 72°C to 74°C
Beilstein 135479
Formula Weight 212.46
Physical Form Powder
Percent Purity ≥99%
Chemical Name or Material 2-(Trichloroacetyl)pyrrole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures