Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(Trifluoromethoxy)benzenesulfonamide, 99%

Click to view available options
Quantity:
1g
5g
37526-59-3
C7H6F3NO3S
241.184
MFCD01320751
HIFGQHGWMTZMOH-UHFFFAOYSA-N
2-trifluoromethoxy benzenesulfonamide,benzenesulfonamide, 2-trifluoromethoxy,unii-3dq25354mb,2-trifluoromethoxy benzenesulphonamide,2-trifluoromethoxy benzene-1-sulfonamide,acmc-209itm,dsstox_cid_24930,dsstox_rid_80595,dsstox_gsid_44930,ksc222c3t
2777208
2-(trifluoromethoxy)benzenesulfonamide
C1=CC=C(C(=C1)OC(F)(F)F)S(=O)(=O)N
37526-59-3
C7H6F3NO3S
241.184
MFCD01320751
HIFGQHGWMTZMOH-UHFFFAOYSA-N
2-trifluoromethoxy benzenesulfonamide,benzenesulfonamide, 2-trifluoromethoxy,unii-3dq25354mb,2-trifluoromethoxy benzenesulphonamide,2-trifluoromethoxy benzene-1-sulfonamide,acmc-209itm,dsstox_cid_24930,dsstox_rid_80595,dsstox_gsid_44930,ksc222c3t
2777208
2-(trifluoromethoxy)benzenesulfonamide
C1=CC=C(C(=C1)OC(F)(F)F)S(=O)(=O)N
Melting Point 187°C to 190°C
Quantity 5g
Formula Weight 241.19
Percent Purity 99%
Chemical Name or Material 2-(Trifluoromethoxy)benzenesulfonamide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning