Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(Trifluoromethoxy)benzoyl chloride, 97+%

Manufacturer:  Alfa Aesar™ 43753-06

Quantity 5g

 View more versions of this product

Catalog No. AA4375306

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

116827-40-8
224.57
UN3094
C8H4ClF3O2
MFCD00052330
2- (Trifluoromethoxy)benzoyl chloride

Specifications

94° to 96°C (15mmHg)
116827-40-8
Liquid
224.57
UN3094
≥97%
5g
1.4700
C8H4ClF3O2
MFCD00052330
2- (Trifluoromethoxy)benzoyl chloride

Safety and Handling

GHS H Statement
H261-H314-H318

GHS P Statement
P231+P232-P260-P280-P303+P361+P353-P305+P351+P338-P301+P330+P331-P304+P340-P310-P405-P501a

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, WATER-REACTIVE, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit