Alfa Aesar™ 2-(Trifluoromethoxy)benzoyl chloride, 97+%

Manufacturer: Alfa Aesar™ 43753-03

Quantity 1g

 View more versions of this product

Catalog No. AA4375303

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 116827-40-8
    Molecular Formula C8H4ClF3O2
    Formula Weight 224.57
    MDL Number MFCD00052330
    UN Number UN3094
    Chemical Name or Material 2- (Trifluoromethoxy)benzoyl chloride

    Specifications

    Formula Weight 224.57
    CAS 116827-40-8
    Molecular Formula C8H4ClF3O2
    Health Hazard 1 Danger
    Quantity 1g
    Refractive Index 1.4700
    Physical Form Liquid
    MDL Number MFCD00052330
    Health Hazard 3 GHS P Statement
    P231+P232-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Handle under inert gas.
    Protect from moisture.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contamin
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, WATER-REACTIVE, N.O.S.
    Boiling Point 94° to 96°C (15mmHg)
    UN Number UN3094
    Health Hazard 2 GHS H Statement
    H261-H314-H318
    In contact with water releases flammable gas.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Chemical Name or Material 2- (Trifluoromethoxy)benzoyl chloride
    Assay ≥97%

    GHS H Statement
    H261-H314-H318
    In contact with water releases flammable gas.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P231+P232-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Handle under inert gas.
    Protect from moisture.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contamin

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit