Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Trifluoromethyl)-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine dihydrochloride, 95%, Acros Organics™

Click to view available options
Quantity:
250mg
1g
1311254-51-9
C7H8F3N3·2ClH
264.08
NWCODVVXJNWZAL-UHFFFAOYSA-N
2-trifluoromethyl-5,6,7,8-tetrahydroimidazo 1,2-a pyrazine dihydrochloride,2-trifluoromethyl-5h,6h,7h,8h-imidazo 1,2-a pyrazine dihydrochloride
56845330
2-(trifluoromethyl)-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine;dihydrochloride
C1CN2C=C(N=C2CN1)C(F)(F)F.Cl.Cl
1311254-51-9
C7H8F3N3·2ClH
264.08
NWCODVVXJNWZAL-UHFFFAOYSA-N
2-trifluoromethyl-5,6,7,8-tetrahydroimidazo 1,2-a pyrazine dihydrochloride,2-trifluoromethyl-5h,6h,7h,8h-imidazo 1,2-a pyrazine dihydrochloride
56845330
2-(trifluoromethyl)-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine;dihydrochloride
C1CN2C=C(N=C2CN1)C(F)(F)F.Cl.Cl
Packaging Glass Bottle
Quantity 1g
Melting Point 223.0°C to 226.0°C
Infrared Spectrum Authentic
Assay Percent Range 94.0 to max. 106% min. (Argentometry)
Formula Weight 264.08
Percent Purity 95%
Chemical Name or Material 2-(Trifluoromethyl)-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine dihydrochloride

GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning