Alfa Aesar™ 2'-(Trifluoromethyl)acetophenone, 97+%

Manufacturer: Alfa Aesar™ A1224803

Quantity 1g

 View more versions of this product

Catalog No. AAA1224803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 17408-14-9
    Molecular Formula C9H7F3O
    Formula Weight 188.15
    MDL Number MFCD00000378
    Beilstein 2259035
    Synonym 2-Acetylbenzotrifluoride
    Chemical Name or Material 2'- (Trifluoromethyl)acetophenone

    Specifications

    Formula Weight 188.15
    CAS 17408-14-9
    Odor Characteristic
    MDL Number MFCD00000378
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Boiling Point 163° to 165°C
    Beilstein 2259035
    Synonym 2-Acetylbenzotrifluoride
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Molecular Formula C9H7F3O
    Flash Point 84°C (183°F)
    Melting Point 15° to 17°C
    Quantity 1g
    Chemical Name or Material 2'- (Trifluoromethyl)acetophenone
    Assay ≥97%

    GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit