Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(Trifluoromethyl)benzoic acid, 98+%

Click to view available options
Quantity:
5g
25g
100g
433-97-6
C8H5F3O2
190.121
MFCD00002476
FBRJYBGLCHWYOE-UHFFFAOYSA-N
2-trifluoromethyl benzoic acid,o-trifluoromethylbenzoic acid,alpha,alpha,alpha-trifluoro-o-toluic acid,o-carboxybenzotrifluoride,2-carboxybenzotrifluoride,unii-0r1ck7mzua,trifluoromethylbenzoic acid,2-trifluoromethyl benzoate,2-trifluromethylbenzoic acid,trifluoromethyl benzoic acid
9899
CHEBI:60694
2-(trifluoromethyl)benzoic acid
C1=CC=C(C(=C1)C(=O)O)C(F)(F)F
433-97-6
C8H5F3O2
190.121
MFCD00002476
FBRJYBGLCHWYOE-UHFFFAOYSA-N
2-trifluoromethyl benzoic acid,o-trifluoromethylbenzoic acid,alpha,alpha,alpha-trifluoro-o-toluic acid,o-carboxybenzotrifluoride,2-carboxybenzotrifluoride,unii-0r1ck7mzua,trifluoromethylbenzoic acid,2-trifluoromethyl benzoate,2-trifluromethylbenzoic acid,trifluoromethyl benzoic acid
9899
CHEBI:60694
2-(trifluoromethyl)benzoic acid
C1=CC=C(C(=C1)C(=O)O)C(F)(F)F
Quantity 25g
Boiling Point 247°C to 254°C
Melting Point 107°C to 110°C
Beilstein 976984
Formula Weight 190.12
Percent Purity ≥98%
Chemical Name or Material 2-(Trifluoromethyl)benzoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 207-093-9

TSCA : Yes