Alfa Aesar™ 2-(Trifluoromethyl)benzoyl chloride, 98%

Manufacturer: Alfa Aesar™ A1056422

Quantity 100g

 View more versions of this product

Catalog No. AAA1056422

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 312-94-7
    Molecular Formula C8H4ClF3O
    Formula Weight 208.57
    MDL Number MFCD00000667
    UN Number UN3265
    Beilstein 881040
    Synonym alpha,alpha,alpha-Trifluoro-o-toluoyl chloride
    Chemical Name or Material 2- (Trifluoromethyl)benzoyl chloride

    Specifications

    Formula Weight 208.57
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Health Hazard 1 Danger
    Molecular Formula C8H4ClF3O
    MDL Number MFCD00000667
    Beilstein 881040
    Boiling Point 89° to 90°C (16mmHg)
    Melting Point −22°C
    Odor Pungent
    UN Number UN3265
    Synonym alpha,alpha,alpha-Trifluoro-o-toluoyl chloride
    CAS 312-94-7
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
    Flash Point 95°C (203°F)
    Quantity 100g
    Chemical Name or Material 2- (Trifluoromethyl)benzoyl chloride
    Assay 98%

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit