Alfa Aesar™ 2-(Trifluoromethyl)phenylacetylene, 97%

Manufacturer: Alfa Aesar™ H5347806

Quantity 5g

 View more versions of this product

Catalog No. AAH5347806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 704-41-6
    Molecular Formula C9H5F3
    Formula Weight 170.13
    MDL Number MFCD00467313
    UN Number UN1993
    Synonym 1-Ethynyl-2-(trifluoromethyl)benzene; 2-Ethynyl-alpha,alpha,alpha-trifluorotoluene
    Chemical Name or Material 2-(Trifluoromethyl)phenylacetylene

    Specifications

    Formula Weight 170.13
    UN Number UN1993
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Quantity 5g
    Molecular Formula C9H5F3
    MDL Number MFCD00467313
    CAS 704-41-6
    Synonym 1-Ethynyl-2-(trifluoromethyl)benzene; 2-Ethynyl-alpha,alpha,alpha-trifluorotoluene
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226
    Flammable liquid and vapor.
    Flash Point 25°C (77°F)
    Boiling Point 45°C (8mmHg)
    Chemical Name or Material 2-(Trifluoromethyl)phenylacetylene
    Assay 97%

    Description

    Applications
    2-(Trifluoromethyl)phenylacetylene is used as pharmaceutical intermediate.

    Solubility
    Sparingly soluble in water.

    Notes
    Stable under recommended storage conditions. Store away from oxidizing agents.

    GHS H Statement
    H226
    Flammable liquid and vapor.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit