Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 2-(Trifluoromethyl)phenylhydrazine hydrochloride, 98%
Click to view available options
Quantity:
5g
25g
100g
Specifications
Sensitivity | Hygroscopic |
Melting Point | ~235°C (decomposition) |
Quantity | 25g |
Beilstein | 4582303 |
Formula Weight | 212.6 |
Percent Purity | 98% |
Chemical Name or Material | 2-(Trifluoromethyl)phenylhydrazine hydrochloride |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
H301+H311+H331-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures