Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(Trifluoromethyl)phenylhydrazine hydrochloride, 98%

Click to view available options
Quantity:
5g
25g
100g
3107-34-4
C7H8ClF3N2
212.6
MFCD00102619
ZUSWDTWYONAOPH-UHFFFAOYSA-N
2-trifluoromethyl phenylhydrazine hydrochloride,2-trifluoromethyl phenylhydrazine hcl,2-hydrazinobenzotrifluoride hcl,2-hydrazinobenzotrifluoride hydrochloride,2-trifluoromethyl phenyl hydrazine hydrochloride,2-trifluoromethyl phenylhydrazine monohydrochloride,1-2-trifluoromethyl phenyl hydrazine hydrochloride,n'-2-trifluoromethyl-phenyl-hydrazinium, chloride,2-trifluoromethylphenylhydrazine hydrochloride
12891828
[2-(trifluoromethyl)phenyl]hydrazine;hydrochloride
C1=CC=C(C(=C1)C(F)(F)F)NN.Cl
3107-34-4
C7H8ClF3N2
212.6
MFCD00102619
ZUSWDTWYONAOPH-UHFFFAOYSA-N
2-trifluoromethyl phenylhydrazine hydrochloride,2-trifluoromethyl phenylhydrazine hcl,2-hydrazinobenzotrifluoride hcl,2-hydrazinobenzotrifluoride hydrochloride,2-trifluoromethyl phenyl hydrazine hydrochloride,2-trifluoromethyl phenylhydrazine monohydrochloride,1-2-trifluoromethyl phenyl hydrazine hydrochloride,n'-2-trifluoromethyl-phenyl-hydrazinium, chloride,2-trifluoromethylphenylhydrazine hydrochloride
12891828
[2-(trifluoromethyl)phenyl]hydrazine;hydrochloride
C1=CC=C(C(=C1)C(F)(F)F)NN.Cl
Melting Point ~235°C (decomposition)
Quantity 100g
Beilstein 4582303
Formula Weight 212.6
Percent Purity 98%
Chemical Name or Material 2-(Trifluoromethyl)phenylhydrazine hydrochloride

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger