Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Trifluoromethyl)pyrazine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
61655-67-2
C5H3F3N2
148.09
MFCD06797734
CEAMSISEJZMQEP-UHFFFAOYSA-N
2-trifluoromethyl pyrazine,2-trifluoromethyl-pyrazine,pyrazine, trifluoromethyl,trifluoromethylpyrazine,pyrazine, 2-trifluoromethyl
17888936
2-(trifluoromethyl)pyrazine
C1=CN=C(C=N1)C(F)(F)F
61655-67-2
C5H3F3N2
148.09
MFCD06797734
CEAMSISEJZMQEP-UHFFFAOYSA-N
2-trifluoromethyl pyrazine,2-trifluoromethyl-pyrazine,pyrazine, trifluoromethyl,trifluoromethylpyrazine,pyrazine, 2-trifluoromethyl
17888936
2-(trifluoromethyl)pyrazine
C1=CN=C(C=N1)C(F)(F)F
Boiling Point 116.0°C to 118.0°C
Color Colorless to Yellow
Infrared Spectrum Authentic
Quantity 1g
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 148.09
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethyl)pyrazine
Show More Show Less

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

GHS Signal Word: Warning