Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Trifluoromethyl)pyrazine, 97%, Thermo Scientific™

Catalog Number AC432610050
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
61655-67-2
C5H3F3N2
148.09
MFCD06797734
CEAMSISEJZMQEP-UHFFFAOYSA-N
2-trifluoromethyl pyrazine, 2-trifluoromethyl-pyrazine, pyrazine, trifluoromethyl, trifluoromethylpyrazine, pyrazine, 2-trifluoromethyl
17888936
2-(trifluoromethyl)pyrazine
C1=CN=C(C=N1)C(F)(F)F
61655-67-2
C5H3F3N2
148.09
MFCD06797734
CEAMSISEJZMQEP-UHFFFAOYSA-N
2-trifluoromethyl pyrazine, 2-trifluoromethyl-pyrazine, pyrazine, trifluoromethyl, trifluoromethylpyrazine, pyrazine, 2-trifluoromethyl
17888936
2-(trifluoromethyl)pyrazine
C1=CN=C(C=N1)C(F)(F)F
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 116.0°C to 118.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 148.09
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-(Trifluoromethyl)pyrazine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit