Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(Trifluoromethyl)thioxanthen-9-one, 98%

Click to view available options
Quantity:
1g
5g
25g
1693-28-3
C14H7F3OS
280.264
MFCD00079785
NEWRXGDGZGIHIS-UHFFFAOYSA-N
2-trifluoromethyl thioxanthen-9-one,2-trifluoromethyl thioxanthone,2-trifluoromethyl-9h-thioxanthen-9-one,2-trifluoromethylthioxanthone,9h-thioxanthen-9-one, 2-trifluoromethyl,2-trifludromethylthioxanthone,2-trifluormethyl-9-thiaxanthen-9-one,2-trifluoromethyl dibenzo b,e thiin-10-one,pubchem10678,acmc-209dzj
74324
2-(trifluoromethyl)thioxanthen-9-one
C1=CC=C2C(=C1)C(=O)C3=C(S2)C=CC(=C3)C(F)(F)F
1693-28-3
C14H7F3OS
280.264
MFCD00079785
NEWRXGDGZGIHIS-UHFFFAOYSA-N
2-trifluoromethyl thioxanthen-9-one,2-trifluoromethyl thioxanthone,2-trifluoromethyl-9h-thioxanthen-9-one,2-trifluoromethylthioxanthone,9h-thioxanthen-9-one, 2-trifluoromethyl,2-trifludromethylthioxanthone,2-trifluormethyl-9-thiaxanthen-9-one,2-trifluoromethyl dibenzo b,e thiin-10-one,pubchem10678,acmc-209dzj
74324
2-(trifluoromethyl)thioxanthen-9-one
C1=CC=C2C(=C1)C(=O)C3=C(S2)C=CC(=C3)C(F)(F)F
Quantity 1g
Melting Point 147°C to 151°C
Color Yellow
Sensitivity Light Sensitive
Beilstein 1583738
Formula Weight 280.26
Physical Form Crystalline
Percent Purity 98%
Chemical Name or Material 2-(Trifluoromethyl)thioxanthen-9-one

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning