Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Trimethylsilyl)ethanol, 99%, ACROS Organics™

Click to view available options
Quantity:
10mL
50mL
250mL
Packaging:
Glass bottle
2916-68-9
C5H14OSi
118.25
MFCD00002825
ZNGINKJHQQQORD-UHFFFAOYSA-N
2-trimethylsilyl ethanol,ethanol, 2-trimethylsilyl,2-hydroxyethyl trimethylsilane,2-trimethylsilyl ethan-1-ol,silane, 2-hydroxyethyl trimethyl,2-trimethylsilanyl-ethanol,2-trimethylsilyl-ethanol,2-trimethylsilyl ethanal
18013
2-trimethylsilylethanol
C[Si](C)(C)CCO
2916-68-9
C5H14OSi
118.25
MFCD00002825
ZNGINKJHQQQORD-UHFFFAOYSA-N
2-trimethylsilyl ethanol,ethanol, 2-trimethylsilyl,2-hydroxyethyl trimethylsilane,2-trimethylsilyl ethan-1-ol,silane, 2-hydroxyethyl trimethyl,2-trimethylsilanyl-ethanol,2-trimethylsilyl-ethanol,2-trimethylsilyl ethanal
18013
2-trimethylsilylethanol
C[Si](C)(C)CCO
Color Colorless
Flash Point 50°C
Packaging Glass bottle
Boiling Point 71.0°C to 73.0°C (35.0mmHg)
Specific Gravity 0.825
Infrared Spectrum Authentic
Density 0.8250g/mL
Quantity 10mL
Refractive Index 1.4235 to 1.4255
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (CH3)3SiCH2CH2OH
Solubility Information Solubility in water: soluble in water
Formula Weight 118.25
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 2-(Trimethylsilyl)ethanol
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 220-844-5