Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Trimethylsilyl-N-tert-butylacetaldimine, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
73198-78-4
C9H21NSi
171.359
MFCD01632577
FBKXKUUGFCDLGG-UHFFFAOYSA-N
2-trimethylsilyl-n-tert-butylacetaldimine,trimethylsilylacetaldehyde tert-butylimine,n-tert-butyl-2-trimethylsilyl ethanimine,tert-butyl 2-trimethylsilyl ethylidene amine,1e-n-tert-butyl-2-trimethylsilyl ethanimine,1z-n-tert-butyl-2-trimethylsilyl ethanimine,e-tert-butyl 2-trimethylsilyl ethylidene amine
2733576
N-tert-butyl-2-trimethylsilylethanimine
CC(C)(C)N=CC[Si](C)(C)C
73198-78-4
C9H21NSi
171.359
MFCD01632577
FBKXKUUGFCDLGG-UHFFFAOYSA-N
2-trimethylsilyl-n-tert-butylacetaldimine,trimethylsilylacetaldehyde tert-butylimine,n-tert-butyl-2-trimethylsilyl ethanimine,tert-butyl 2-trimethylsilyl ethylidene amine,1e-n-tert-butyl-2-trimethylsilyl ethanimine,1z-n-tert-butyl-2-trimethylsilyl ethanimine,e-tert-butyl 2-trimethylsilyl ethylidene amine
2733576
N-tert-butyl-2-trimethylsilylethanimine
CC(C)(C)N=CC[Si](C)(C)C
Quantity 1g
Packaging Glass bottle
Refractive Index 1.4250 to 1.4290 (20°C, 589nm)
Color Yellow
Infrared Spectrum Authentic
NMR Complies
Linear Formula (CH3)3SiCHCHNC(CH3)3
Formula Weight 171.36
Physical Form Liquid
Chemical Name or Material 2-Trimethylsilyl-N-tert-butylacetaldimine