Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Trimethylsilyl)pyridine +97%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
Packaging:
Glass bottle
13737-04-7
C8H13NSi
151.284
MFCD06797181
IBLKFJNZUSUTPB-UHFFFAOYSA-N
2-trimethylsilyl pyridine,pyridine, 2-trimethylsilyl,2-pyridyltrimethylsilane,2-trimethylsilylpyridine,2-pyridyl trimethylsilane,trimethyl pyridin-2-yl silane,amtsi028,acmc-1cb48,trimethyl 2-pyridinyl silane
139573
trimethyl(pyridin-2-yl)silane
C[Si](C)(C)C1=CC=CC=N1
13737-04-7
C8H13NSi
151.284
MFCD06797181
IBLKFJNZUSUTPB-UHFFFAOYSA-N
2-trimethylsilyl pyridine,pyridine, 2-trimethylsilyl,2-pyridyltrimethylsilane,2-trimethylsilylpyridine,2-pyridyl trimethylsilane,trimethyl pyridin-2-yl silane,amtsi028,acmc-1cb48,trimethyl 2-pyridinyl silane
139573
trimethyl(pyridin-2-yl)silane
C[Si](C)(C)C1=CC=CC=N1
Color Colorless to Brown
Quantity 1g
Density 0.911g/mL
Refractive Index 1.4900 to 1.4920 (20°C, 589nm)
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 97.0
CAS Max % 100.0
Assay Percent Range 97+%
Formula Weight 151.28
Physical Form Liquid
Percent Purity ≥97.0%
Chemical Name or Material 2-(Trimethylsilyl)pyridine
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning