Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(Trimethylsilyl)thiazole, 97%

Click to view available options
Quantity:
1g
5g
25g
79265-30-8
C6H11NSSi
157.306
MFCD00066274
VJCHUDDPWPQOLH-UHFFFAOYSA-N
2-trimethylsilyl thiazole,2-trimethylsilylthiazole,2-trimethylsilyl-1,3-thiazole,thiazole, 2-trimethylsilyl,2-trimethylsilanylthiazole,trimethyl 1,3-thiazol-2-yl silane,2-tms-thiazole,pubchem10262,acmc-209pgq
588453
trimethyl(1,3-thiazol-2-yl)silane
C[Si](C)(C)C1=NC=CS1
79265-30-8
C6H11NSSi
157.306
MFCD00066274
VJCHUDDPWPQOLH-UHFFFAOYSA-N
2-trimethylsilyl thiazole,2-trimethylsilylthiazole,2-trimethylsilyl-1,3-thiazole,thiazole, 2-trimethylsilyl,2-trimethylsilanylthiazole,trimethyl 1,3-thiazol-2-yl silane,2-tms-thiazole,pubchem10262,acmc-209pgq
588453
trimethyl(1,3-thiazol-2-yl)silane
C[Si](C)(C)C1=NC=CS1
Refractive Index 1.497
Density 0.985
Sensitivity Moisture sensitive
Quantity 1g
Boiling Point 53°C to 55°C (10mmHg)
Flash Point 57°C (135°F)
UN Number UN1993
Formula Weight 157.31
Percent Purity 97%
Chemical Name or Material 2-(Trimethylsilyl)thiazole

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P280-P370+P378q-P501c

H227

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : No

Recommended Storage : Keep cold