Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Undecanone, 98%, Acros Organics™

Click to view available options
Quantity:
5mL
250mL
1L
Packaging:
Glass bottle
112-12-9
C11H22O
170.3
MFCD00009583
KYWIYKKSMDLRDC-UHFFFAOYSA-N
2-undecanone,methyl nonyl ketone,2-hendecanone,ketone, methyl nonyl,undecanone,nonyl methyl ketone,rue ketone,2-oxoundecane,methyl n-nonyl ketone,methylnonylketone
8163
CHEBI:17700
undecan-2-one
CCCCCCCCCC(=O)C
112-12-9
C11H22O
170.3
MFCD00009583
KYWIYKKSMDLRDC-UHFFFAOYSA-N
2-undecanone,methyl nonyl ketone,2-hendecanone,ketone, methyl nonyl,undecanone,nonyl methyl ketone,rue ketone,2-oxoundecane,methyl n-nonyl ketone,methylnonylketone
8163
CHEBI:17700
undecan-2-one
CCCCCCCCCC(=O)C
Density 0.8240g/mL
Boiling Point 231.0°C to 232.0°C
Packaging Glass bottle
Flash Point 88°C
Melting Point 11.0°C to 13.0°C
Infrared Spectrum Authentic
Specific Gravity 0.824
Quantity 1L
Refractive Index 1.4280 to 1.4310
Color Colorless to Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3(CH2)8COCH3
Beilstein 01, 713
Merck Index 15, 6177
Solubility Information Solubility in water: insoluble
Formula Weight 170.3
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2-Undecanone, 99%
Show More Show Less

GHS H Statement:
Toxic to aquatic life with long lasting effects.

GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 203-937-5

RTECSNumber : YQ2820000

TSCA : TSCA