Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Valerylthiophene, 97%

Manufacturer:  Alfa Aesar™ H2632603

Quantity 1g

 View more versions of this product

Catalog No. AAH2632603


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

53119-25-8
168.25
2-Pentanoylthiophene; 1-(2-Thienyl)-1-pentanone
C9H12OS
MFCD00053149
2-Valerylthiophene

Specifications

53119-25-8
168.25
2-Pentanoylthiophene; 1-(2-Thienyl)-1-pentanone
2-Valerylthiophene
C9H12OS
MFCD00053149
1g
97%
missing translation for 'provideContentCorrection'

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

missing translation for 'productTitle'