Alfa Aesar™ 2-Valerylthiophene, 97%

Manufacturer: Alfa Aesar™ H2632609

Quantity 10g

 View more versions of this product

Catalog No. AAH2632609

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 53119-25-8
    Molecular Formula C9H12OS
    Formula Weight 168.25
    MDL Number MFCD00053149
    Synonym 2-Pentanoylthiophene; 1-(2-Thienyl)-1-pentanone
    Chemical Name or Material 2-Valerylthiophene

    Specifications

    Formula Weight 168.25
    Molecular Formula C9H12OS
    MDL Number MFCD00053149
    CAS 53119-25-8
    Synonym 2-Pentanoylthiophene; 1-(2-Thienyl)-1-pentanone
    Quantity 10g
    Chemical Name or Material 2-Valerylthiophene
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit