Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Vinylbenzoic acid, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
As intermediate in orgaic syntheses. Phthalides were obtained in low yields by palladium-assisted cyclization of 2-vinylbenzoic acid and 2-(2-methyl-2-propenyl)- benzoic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
As intermediate in orgaic syntheses. Phthalides were obtained in low yields by palladium-assisted cyclization of 2-vinylbenzoic acid and 2-(2-methyl-2-propenyl)- benzoic acid.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
As intermediate in orgaic syntheses. Phthalides were obtained in low yields by palladium-assisted cyclization of 2-vinylbenzoic acid and 2-(2-methyl-2-propenyl)- benzoic acid.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
Specifications
Melting Point | 94°C |
Quantity | 1 g |
Synonym | 2-vinylbenzoic acid, vinylbenzoic acid, benzoic acid, ethenyl, benzoic acid, 2-ethenyl, pubchem19668, benzoic acid,2-ethenyl, benzoic acid, 2-ethenyl-9ci |
Formula Weight | 148.16 |
Percent Purity | 96% |
Chemical Name or Material | 2-Vinylbenzoic acid |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
TSCA | No |
Recommended Storage | Keep cold |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction