Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Vinylnaphthalene, Stabilized 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
827-54-3
C12H10
154.21
MFCD00004125
KXYAVSFOJVUIHT-UHFFFAOYSA-N
2-vinylnaphthalene,naphthalene, 2-ethenyl,2-vinyl-naphthalene,beta-vinylnaphthalene,poly 2-vinylnaphthalene,unii-hzd8li91n1,hzd8li91n1,polyvinylnaphthalene,2-vinyl naphthalene,2-ethenyinaphthalene
13230
CHEBI:51325
2-ethenylnaphthalene
C=CC1=CC2=CC=CC=C2C=C1
827-54-3
C12H10
154.21
MFCD00004125
KXYAVSFOJVUIHT-UHFFFAOYSA-N
2-vinylnaphthalene,naphthalene, 2-ethenyl,2-vinyl-naphthalene,beta-vinylnaphthalene,poly 2-vinylnaphthalene,unii-hzd8li91n1,hzd8li91n1,polyvinylnaphthalene,2-vinyl naphthalene,2-ethenyinaphthalene
13230
CHEBI:51325
2-ethenylnaphthalene
C=CC1=CC2=CC=CC=C2C=C1
Packaging Glass bottle
Color Beige to White or Yellow
Melting Point 63°C to 67°C
Boiling Point 135°C (18.0mmHg)
Infrared Spectrum Authentic
Quantity 25g
Stabilizer 0.5% max. p-tert.-butylcatechol
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula C10H7CH=CH2
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,acetone,benzene
Formula Weight 154.21
Physical Form Powder
Percent Purity ≥96.0%
Chemical Name or Material 2-Vinylnaphthalene, 98%
Show More Show Less

EINECSNumber : 212-573-6

TSCA : TSCA