Learn More
Medchemexpress LLC 20(S)-hydroxycholesterol | 516-72-3 | MFCD16661190 | 99.3% | 402.65 | C27H46O2 | 10MM 1ML
SDP
Supplier:  Medchemexpress LLC HY1231610MM 1ML
Encompass_Preferred
20(S)-Hydroxycholesterol is an oxysterol that functions as an allosteric activator of the Smoothened (Smo) receptor in the Hedgehog signaling pathway. It is supplied as a powder and as a ready-to-use 10 mM solution in DMSO (1 mL) for research applications in developmental biology, oncology, bone regeneration, and angiogenesis.
  • Allosteric activator of Smoothened for Hedgehog pathway studies.
  • Available as powder and as a 10 mM solution in DMSO (1 mL).
  • High purity suitable for analytical and research use.
  • Documented molecular weight and formula for experimental planning.
  • Recommended storage conditions provided for powder and solvent solutions.
Catalog No. 50-002-35615