Learn More
Invitrogen™ Varicella zoster Virus glycoprotein E Recombinant Rabbit Monoclonal Antibody (276)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA560398

Description
Varicella zoster Virus glycoprotein E Recombinant Monoclonal Antibody for Western Blot, ELISA
Varicella Zoster Virus (VZV), a member of the human herpes virus family, causes two distinct clinical manifestations: childhood chickenpox(Varicella) and shingles (zoster). Varicella is the outcome of the primary infection with VZV, whereas, zoster is the result of VZV reactivation from latently infected sensory ganglia which occurs predominantly in aging and immunosuppressed individuals' virons are spherical and 150-200 nm in diameter. Its lipid envelope encloses the nucleocapsid of 162 capsomeres arranged in a hexagonal form. Its DNA is a single linear, double strand molecule, 125,000 nt long.
Specifications
Varicella zoster Virus glycoprotein E | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Recombinant Varicella-zoster virus (VZV)/HHV-3 gE/glycoprotein E protein (Met1-Tyr538). | |
20 μg | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
276 | |
Unconjugated | |
Rabbit | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.