Learn More
Greiner Bio-One™ 200µL; Pipette Tips for Gilson™ Pipetman Pipettes
Provides optimum performance and fit for Gilson Pipetman Pipettes.
Specifications
Volume (Metric) 10 to 200μL
Certifications/Compliance ISO 9001
Tip Style Extended Length
Graduated No
For Use With (Equipment) Gilson™ Pipetman™ F10
View More Specs
Products 2
Catalog Number Mfr. No. Color Sterility Material Price Quantity & Availability  
Catalog Number Mfr. No. Color Sterility Material Price Quantity & Availability  
07-000-816 View Documents Greiner Bio-One™
739261
Yellow Sterile PP (Polypropylene) This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
07-000-820 View Documents Greiner Bio-One™
739290
Yellow Non-sterile PE (Polyethylene) This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
  • Manufactured from high-grade polypropylene
  • Polypropylene has high breaking strength, dimensionally and stable heat-resistant
  • Free of heavy metals
  • Manufactured under production conditions certified conforming to ISO 9001 sta
Specifications
Specifications
10 to 200μL
Extended Length
Gilson™ Pipetman™ F10
Yes
ISO 9001
No
Yellow
−190° to 140°C