Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

200 Proof Alcohol for Extraction CDA-12A (Ethanol and n-Heptane), Reagents

Catalog No. p-200021228
Click to view available options
Quantity:
20 L
55 gal.
270 gal.

200 Proof Alcohol for Extraction CDA-12A (Ethanol and n-Heptane), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64-17-5, 142-82-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym Ethanol, Ethyl alcohol, Methylcarbinol
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -55°C
Color Colorless
Boiling Point 78°C
Quantity 20 L
Packaging Steel Pail
Physical Form Liquid
Specific Gravity 0.7974
Percent Purity Min. 99.5%
Grade Reagent
Chemical Name or Material 200 Proof Alcohol for Extraction CDA-12A (Ethanol + n-Heptane)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.