Learn More
Thermo Scientific™ 20mm Headspace Vials & Closures Available on GSA/VA Contract for Federal Government customers only.
Save time and reduce risk of contamination. These convenience kits offer matched quantities of 20mm headspace vials.
Manufacturer:  Thermo Scientific™ 60180519
 View more versions of this product
Catalog No. 03-251-413
  Check Availability
Add to cart
Description
Description
Save time during sample preparation and reduce the risk of contamination using Thermo Scientific™ 20mm Crimp Top Clear Headspace Vial Convenience Kits (Unassembled). Each kit includes matched quantities of vials and caps with pre-assembled septa. Choose from Thermo Scientific™ National™ and Thermo Scientific™ Chromacol™ brands.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Crimp Cap
20mm Composite Magnetic Crimp Cap, 8mm hole
100
Alu/Tinplate
20mm
Aluminum/Tin Plate
Blue
Crimp Cap (Magnetic, Composite)
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.