Learn More
US Plastic Corporation 22-Ounce HDPE Spray Bottle, Oval, Blue and White Sprayer
SDP
Supplier:  US Plastic Corporation 66217
Encompass
Spray Bottle with Sprayers, 22 oz. Bottle with 28mm Blue and White Sprayer, high Density Polyethylene Bottles and Polypropylene Sprayer for Waxes, Starches, Cleaners, Disinfectants, Air Fresheners, Deodorants, Insecticides
Catalog No. NC9262770