Learn More
Boekel Scientific™ Tissue Cassette Storage Cabinet
Boekel Scientific™ Tissue Cassette Storage Cabinet is made of an epoxy painted cold rolled steel and has the capacity to store up to 1,200 cassettes. Additional Tray Inserts sold separately.
Supplier: Boekel Scientific™ 143000
Description
- Store up to 1,200 Tissue Cassettes
- Stack up to three units high
- Non-tip design
- Each drawer includes a clear tray and foam organizers
Specifications
Light Brown | |
Epoxy Painted Metal | |
7.75 in. | |
3 lb. | |
13.25 in. |
1200 Tissue Cassettes | |
One Clear Plastic Tray per Shelf, Customizable Labels | |
19.7 cm | |
1.4 kg | |
33.7 cm |