Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DenLine Uniforms Protection Plus II Round Neck Unisex 31.5 in. (80 cm) Short Lab Jacket- Fluid Resistant Barrier (AAMI Level 1) with High Air permeability and Comfort
Click to view available options
Color:
Black
Royal Blue
White
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
Primary Applications
· Anatomic Pathology · Clinical Pathology · Clinical Laboratory · Phlebotomy · Specimen Collection · Blood Collection, Testing, & Manufacturing · Human & Animal Research · Academia · Dialysis · Dental · Clean Room ISO 5/Class 100 - Medical Device, Pharma, Electronics
Wearer Protection Provided
- AAMI PB70 Level 1 Protection against Blood & OPIM; Front, Sleeves, Shoulders and Back
- Provides wearers 5 ft. 2 in. and shorter frontal coverage to Knees for OSHA 29 CFR 1910 compliance.
- Provides all height wearers protection to ankles when worn in combination with NEW DenLine style DL385 UltraLite Unisex Drawstring Pant (see DL385 specification information).
Wearer Comfort
- Overall Air Permeability 45 CFM (ASTM D737)
- Experience Industry Leading Comfort & Breathability in a Fluid Resistant Short Jacket
Other Features
- Round Neck made of jacket fabric…no knit ribbing to soil, fray or wear out prematurely
- Stain Resistant Fabric and Cuffs (blood, bleach and ALL aqueous reagents & liquids)
- Anti-static Fabric prevents false instrument readings, damage to computers.
- Lint Free Fabric prevents sample contamination, ideal for molecular testing
- Durable Construction
- 3 Pockets
- Stainless Snap Closures – Lifetime Guaranteed
- Durable Cuffs for Overgloving – Bleach Safe all colors
- Choice of 4 Colors White, Ceil Blue, Royal and Black; Sizes XS to 5XL
- Autoclavable Fabric (4 Min @ 274°F - 134.4°C)
- 225 Commercial Wash Tested; Low Lifetime coat per wash.
Launder using Home/Laundromat or Commercial Equipment; Instructions furnished meet CDC/HICPAC Recommendations for processing medical bloodborne Linen.
Specifications
Type | Lab Coat |
Size | Medium |
Gender | Unisex |
Material | Textured Polyester, Carbon |
Color | Royal Blue |
Closure Type | Snaps |
Cuff Style | Knit |
Disposable | Reusable |
Sterility | Non-sterile |
Resistance | AAMI PB70 Level 1 |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction