Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ 22q13.1/CEN22 FISH Probe

Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.

$2230.00

Specifications

Description DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
Includes DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
For Use With (Application) FISH-Ce
Format Liquid
Storage Requirements Store at 4°C in the dark.
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-932-018
View Documents
Abnova™
FG0146
100 µL
Each for $2,230.00
Add to cart
 
Description

Description

Specifications

Specifications

DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
FISH-Ce
Store at 4°C in the dark.
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
Liquid
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit