Learn More
COPAN Universal Transport Medium
Universal Transport Medium, is a room temperature, stable, viral transport medium for collection, transport, maintenance and long term freeze storage of Viruses, such as Influenza H1N1, Chlamydia, Mycoplasma and Ureaplasma specimens.
Manufacturer:  COPAN 3C036NHL
 View more versions of this product
Catalog No. 23-001-720
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product. View substitute products »
This item is not returnable. View return policy
Description
Description
  • Universal Room Temperature Stable System
  • Used successfully for Rapid Antigen Testing, DFA, Viral Culture and for Molecular-based Assays.
  • Prior to use, store at room temperature
  • Selection of Tube Sizes and Collection Applicators Available
  • Available in bulk or as patient collection packs, which incorporate different combinations of Polyester swabs, as well as mini tip or regular flocked swabs (FLOQSwabs™) and media tubes to suit different work patterns and preferences.
  • Safe and Reliable Molded Breakpoint Swab
    • All swabs feature either a pre-scored breakpoint (polyester swabs) or a unique molded breakpoint (flocked swabs).
    • Pre-scored and molded breakpoints feature allows for safe, reliable breaking of the swab into transport tube.
  • Shatterproof skirted tubes with distinctive internal conical shape enable centrifugation of samples and allows the tubes to stand upright on the laboratory bench.
  • Three glass beads in each tube facilitate the release and dispersion of patient sample material and virus particles from the swab during vortexing.
  • UTM is identical to BD UVT
Specifications
Specifications
Clear
UTM
Sterile
300/Pk.
Screw Cap
16 mm
UTM with Flexible minitip FLOQSwab; Compatible with Roche Liat Flu A/B, RSV
3 mL
100 mm