Learn More
Copan Diagnostics™ UriSponge™: Urine Specimen Collection, Preservation and Transport System
Allows safe collection and preservation of urine specimens without the added expense and hazard of hypodermic urine transfer straws
Supplier: Copan Diagnostics™ 8C023S01.A
Description
- Urine collection device combines a plastic tube and a screw cap with attached sponges impregnated with boric acid and sodium formate urine preservatives
- Maintains viability of bacteria at refrigerated and room temperature for up to 48 hours
- No hypodermic needle urine transfer straw needed, eliminating potential injury and sharps disposal costs
- Reduce repeat collections from patient due to inaccurate filling of boric acid tubes
- Automatically processed on WASP™ using its built-in spinner, which easily elutes urine from the sponges prior to planting and streaking
- If processed manually, simply centrifuge the tube to release urine from the sponge
Specifications
Foam Sponge Urine Collection Device | |
Polypropylene with Polyurethane Foam Sponge | |
Individual Peel Pouch |
Screw Cap | |
500/Cs. | |
12 x 80 mm |