Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Parter Medical Products™ 24-Hour Urine Containers

Proprietary, leak-resistant closure system assures specimen integrity during collection and transport to the laboratory

Supplier:  Parter Medical Products™ 126912

 View more versions of this product

Catalog No. 22-150-115

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

Explore available promotions

Description

Description

  • Ergonomic Design
  • Balanced CG
  • Capacity: 3.5L
  • EZ-UP Spout Cap (optional)
  • Manufactured from pure virgin, UV filtering, medical grade HDPE
  • Choice of integrated 30mL 6N HCl or none (Value style)

Specifications

Specifications

24-Hour Urine Specimen Collection Container
Yes
Non-sterile
Amber
Used by both the patients and the health care professional for collection and transport of specimen to the laboratory
30/Cs.
EZ-UP Spout Cap
HDPE
70 mm
Horizontal Space Saver
Natural, 70 mm leak resistant HDPE closure with EZ-UP pour spout, Container includes integrated 30 mL 6N HCL
3.5 L
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.